2-Chloro-3,6-difluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005596-5G 005596-1G
FLUO005596-5GEA 0 GBP
FLUO005596-5G FLUO005596-1G
2-Chloro-3,6-difluorobenzyl alcohol
2-Chloro-3,6-difluorobenzyl alcohol
Formula: C₇H₅ClF₂O
MDL Number: MFCD01631445
CAS Number: 261762-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR