2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

009080-5G 009080-25G
FLUO009080-5GEA 0 GBP
FLUO009080-5G FLUO009080-25G
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Formula: C₇H₂ClF₃N₂O₄
Melting Pt: 60…65 °C
MDL Number: MFCD00007076
CAS Number: 392-95-0
EINECS: 206-883-0
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR