3,4-Dichloro-5-nitrobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009130-1G
009130-5G
FLUO009130-1GEA
0
GBP
FLUO009130-1G
FLUO009130-5G
3,4-Dichloro-5-nitrobenzotrifluoride
3,4-Dichloro-5-nitrobenzotrifluoride
Formula:
C₇H₂Cl₂F₃NO₂ |
MDL Number:
MFCD01862035 CAS Number: 657-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...