4-Chloro-3-(trifluoromethyl)aniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002681-100G
002681-500G
FLUO002681-100GEA
0
GBP
FLUO002681-100G
FLUO002681-500G
4-Chloro-3-(trifluoromethyl)aniline
4-Chloro-3-(trifluoromethyl)aniline
Formula:
C₇H₅ClF₃N Boiling Pt: 130…132 °C (27 mmHg) Melting Pt: 36…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007846 CAS Number: 320-51-4 EINECS: 206-277-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...