2-Chloro-6-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-3-chloro-α,α,α-trifluortoluene, 2-Amino-3-chloro-α,α,α-trifluorotoluene, 6-Chloro-α,α,α-trifluoro-o-toluidine, 2-Amino-3-chlorobenzotrifluoride, 2-Chloro-6-trifluoromethylaniline

010358-25G 010358-100G 010358-250G
FLUO010358-25GEA 0 GBP
FLUO010358-25G FLUO010358-100G FLUO010358-250G
2-Chloro-6-(trifluoromethyl)aniline
2-Chloro-6-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
MDL Number: MFCD00272565
CAS Number: 433-94-3
EINECS: 207-091-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR