2-Chloro-5-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Amino-4-chloro-α,α,α-trifluortoluene, 3-Amino-4-chloro-α,α,α-trifluorotoluene, 6-Chloro-α,α,α-trifluoro-m-toluidine, 3-Amino-4-chlorobenzotrifluoride

003964-100G 003964-500G
FLUO003964-100GEA 0 GBP
FLUO003964-100G FLUO003964-500G
2-Chloro-5-(trifluoromethyl)aniline
2-Chloro-5-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 10 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007673
CAS Number: 121-50-6
EINECS: 204-475-7
UN: 2810
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR