2,6-Dichloro-4-(trifluoromethyl)aniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Amino-3,5-dichloro-α,α,α-trifluorotoluene, 4-Amino-3,5-dichlorobenzotrifluoride, 2,6-Dichloro-4-trifluoromethylaniline, 2,6-Dichloro-4-(trifluoromethyl)aniline
CFF548.5
004741-50G
004741-250G
CHMPCFF548.5EA
0
GBP
CHMPCFF548.5
FLUO004741-50G
FLUO004741-250G
2,6-Dichloro-4-(trifluoromethyl)aniline
2,6-Dichloro-4-(trifluoromethyl)aniline
Formula:
C₇H₄Cl₂F₃N MW: 230.02 g/mol Boiling Pt: 60…62 °C (1 mmHg) Melting Pt: 32…35 °C Density: 1.532 g/cm³ (20 °C) |
MDL Number:
MFCD00052918 CAS Number: 24279-39-8 EINECS: 416-430-0 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...