2-Chloro-5-(trifluoromethyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007838-5G 007838-1G
FLUO007838-5GEA 0 GBP
FLUO007838-5G FLUO007838-1G
2-Chloro-5-(trifluoromethyl)benzoic acid
2-Chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
MDL Number: MFCD00061094
CAS Number: 657-06-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR