2-Chloro-6-fluoro-3-methylbenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005644-5G 005644-1G
FLUO005644-5GEA 0 GBP
FLUO005644-5G FLUO005644-1G
2-Chloro-6-fluoro-3-methylbenzamide
2-Chloro-6-fluoro-3-methylbenzamide
Formula: C₈H₇ClFNO
MDL Number: MFCD01631394
CAS Number: 286474-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR