2',4'-Dichloro-5'-fluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019457-25G 019457-100G
FLUO019457-25GEA 0 GBP
FLUO019457-25G FLUO019457-100G
2',4'-Dichloro-5'-fluoroacetophenone
2',4'-Dichloro-5'-fluoroacetophenone
Formula: C₈H₅Cl₂FO
MDL Number: MFCD00077499
CAS Number: 704-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR