2,5-Dichloro-α,α,α-trifluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,4-Dichloro-2-(trifluoromethyl)benzene, 2,5-Dichlorobenzotrifluoride

002689-5G 002689-25G
FLUO002689-5GEA 0 GBP
FLUO002689-5G FLUO002689-25G
2,5-Dichloro-α,α,α-trifluorotoluene
2,5-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Boiling Pt: 181
Melting Pt: 11
Density: 1.5009 g/cm³ (20 °C)
MDL Number: MFCD00000608
CAS Number: 320-50-3
EINECS: 206-276-0
UN: 1760
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR