3,5-Dichloro-α,α,α-trifluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009127-1G
009127-5G
FLUO009127-1GEA
0
GBP
FLUO009127-1G
FLUO009127-5G
3,5-Dichloro-α,α,α-trifluorotoluene
3,5-Dichloro-α,α,α-trifluorotoluene
Formula:
C₇H₃Cl₂F₃ MW: 215 g/mol |
MDL Number:
MFCD00203929 CAS Number: 54773-20-5 EINECS: 259-337-9 UN: 2234 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...