2,3-Dichloro-α,α,α-trifluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004692-1G 004692-5G
FLUO004692-1GEA 0 GBP
FLUO004692-1G FLUO004692-5G
2,3-Dichloro-α,α,α-trifluorotoluene
2,3-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
MDL Number: MFCD00069115
CAS Number: 54773-19-2
EINECS: 259-336-3
UN: 2234
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR