4-Chloro-o-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018393-1G 018393-25G 018393-5G
FLUO018393-1GEA 0 GBP
FLUO018393-1G FLUO018393-25G FLUO018393-5G
4-Chloro-o-toluic acid
4-Chloro-o-toluic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00045852
CAS Number: 7499-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR