5-Chloro-2-methylbenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018395-5G
018395-1G
FLUO018395-5GEA
0
GBP
FLUO018395-5G
FLUO018395-1G
5-Chloro-2-methylbenzoic acid
5-Chloro-2-methylbenzoic acid
Formula:
C₈H₇ClO₂ |
MDL Number:
MFCD00045842 CAS Number: 7499-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...