2',4'-Dichloropropiophenone ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2,4-Dichlorophenyl)-1-propanone, 1-(2,4-Dichlorophenyl)propan-1-one

A11252.22 A11252.06
A11252.22EA 0 GBP
A11252.22 A11252.06
2',4'-Dichloropropiophenone ≥97%
2',4'-Dichloropropiophenone

Purity: 97% CAS No.: 37885-41-9 Molecular Formula: C9H8Cl2O Formula Weight: 203.07 Flash Point: 110°(230°F) Refractive Index: 1.5520 Boiling Point: 121-123°/7mm Beilstein Reference: 2248270 MDL No.: MFCD00027396

Formula: C₉H₈Cl₂O
MW: 203.07 g/mol
Boiling Pt: 225 °C (1013 hPa)
Density: 1.2871 g/cm³ (25 °C)
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00027396
CAS Number: 37885-41-9
EINECS: 253-700-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR