(2,6-Dichlorophenyl)acetonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005453-25G
005453-100G
005453-500G
FLUO005453-25GEA
0
GBP
FLUO005453-25G
FLUO005453-100G
FLUO005453-500G
(2,6-Dichlorophenyl)acetonitrile
(2,6-Dichlorophenyl)acetonitrile
Formula:
C₈H₅Cl₂N Boiling Pt: 109…110 °C (0.4 mmHg) Melting Pt: 75…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001901 CAS Number: 3215-64-3 EINECS: 221-730-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...