4-Tolualdehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: p-Tolualdehyde, p-Tolylaldehyde, p-Methylbenzaldehyd, 4-Methylbenzaldehyd, 4-Methylbenzaldehyde, 4-Tolylaldehyde

M-Clarity™ quality level = MQ300

W306800-10KG-K W306800-1KG-K W306800-25KG-K
SIALW306800-10KG-KEA 428 GBP
SIALW306800-10KG-K SIALW306800-1KG-K SIALW306800-25KG-K
4-Tolualdehyde, Sigma-Aldrich®
4-Tolualdehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006954
CAS Number: 104-87-0
EINECS: 203-246-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR