4-Bromo-o-cresol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Bromo-2-methylphenol, 1-Bromo-4-hydroxy-3-methylbenzene

018299-5G 018299-100G 018299-25G
FLUO018299-5GEA 0 GBP
FLUO018299-5G FLUO018299-100G FLUO018299-25G
4-Bromo-o-cresol
4-Bromo-o-cresol
Formula: C₇H₇BrO
MW: 187.04 g/mol
MDL Number: MFCD00055435
CAS Number: 2362-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR