4-Bromo-m-cresol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018300-10G 018300-100G 018300-25G
FLUO018300-10GEA 0 GBP
FLUO018300-10G FLUO018300-100G FLUO018300-25G
4-Bromo-m-cresol
4-Bromo-m-cresol
Formula: C₇H₇BrO
Boiling Pt: 142…145 °C (23 mmHg)
Melting Pt: 63 °C
MDL Number: MFCD00079723
CAS Number: 14472-14-1
EINECS: 238-464-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR