4-Bromobenzyl alcohol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Bromophenyl)methanol

M-Clarity™ quality level = MQ100

187054-10G 187054-50G
SIAL187054-10GEA 95.8 GBP
SIAL187054-10G SIAL187054-50G
4-Bromobenzyl alcohol, Sigma-Aldrich®
4-Bromobenzyl alcohol
Formula: C₇H₇BrO
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 77…78 °C
MDL Number: MFCD00004650
CAS Number: 873-75-6
EINECS: 212-851-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR