3-Bromobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Bromophenyl)methanol

001365-25G 001365-100G 001365-500G
FLUO001365-25GEA 0 GBP
FLUO001365-25G FLUO001365-100G FLUO001365-500G
3-Bromobenzyl alcohol
3-Bromobenzyl alcohol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 165
Density: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00004629
CAS Number: 15852-73-0
EINECS: 239-975-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR