2-Bromobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Bromophenyl)methanol

008821-10G 008821-25G
FLUO008821-10GEA 0 GBP
FLUO008821-10G FLUO008821-25G
2-Bromobenzyl alcohol
2-Bromobenzyl alcohol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 255
Melting Pt: 80…82 °C
MDL Number: MFCD00004600
CAS Number: 18982-54-2
EINECS: 242-719-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR