2-Bromo-p-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Bromo-4-methylaniline, 4-Amino-3-bromotoluene

001127-25G 001127-100G
FLUO001127-25GEA 0 GBP
FLUO001127-25G FLUO001127-100G
2-Bromo-p-toluidine
2-Bromo-p-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 240
Melting Pt: 26
Density: 1.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007635
CAS Number: 583-68-6
EINECS: 209-515-7
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR