2-Bromo-4-isopropylaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018287-5G 018287-25G
FLUO018287-5GEA 0 GBP
FLUO018287-5G FLUO018287-25G
2-Bromo-4-isopropylaniline
2-Bromo-4-isopropylaniline
Formula: C₉H₁₂BrN
MW: 214.1 g/mol
Boiling Pt: 139…141 °C (20 mmHg)
Density: 1.35 g/cm³ (20 °C)
MDL Number: MFCD00051585
CAS Number: 51605-97-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR