2,6-Dibromo-4-isopropylaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

023322-25G 023322-5G
FLUO023322-25GEA 0 GBP
FLUO023322-25G FLUO023322-5G
2,6-Dibromo-4-isopropylaniline
2,6-Dibromo-4-isopropylaniline
Formula: C₉H₁₁Br₂N
MW: 293 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 45…47 °C
MDL Number: MFCD00015475
CAS Number: 10546-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR