5-Bromo-2-iodotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018284-10G 018284-100G 018284-25G
FLUO018284-10GEA 0 GBP
FLUO018284-10G FLUO018284-100G FLUO018284-25G
5-Bromo-2-iodotoluene
5-Bromo-2-iodotoluene
Formula: C₇H₆BrI
MDL Number: MFCD00060664
CAS Number: 116632-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR