α-Bromo-4-(trifluoromethoxy)toluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-(Bromomethyl)-4-(trifluoromethoxy)benzene, p-(Trifluoromethoxy)benzyl bromide, 4-(Trifluoromethoxy)benzyl bromide

FL8720.1 008720-5G FL8720.10 008720-25G 008720-100G
CHMPFL8720.1EA 0 GBP
CHMPFL8720.1 FLUO008720-5G CHMPFL8720.10 FLUO008720-25G FLUO008720-100G
α-Bromo-4-(trifluoromethoxy)toluene
α-Bromo-4-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 23…24 °C
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00061238
CAS Number: 50824-05-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR