α-Bromo-4-(difluoromethoxy)toluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
004834-5G
004834-1G
FLUO004834-5GEA
0
GBP
FLUO004834-5G
FLUO004834-1G
α-Bromo-4-(difluoromethoxy)toluene
α-Bromo-4-(difluoromethoxy)toluene
Formula:
C₈H₇BrF₂O |
MDL Number:
MFCD00085007 CAS Number: 3447-53-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...