α-Bromo-3-(trifluoromethoxy)toluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001406-1G
001406-5G
FLUO001406-1GEA
0
GBP
FLUO001406-1G
FLUO001406-5G
α-Bromo-3-(trifluoromethoxy)toluene
α-Bromo-e-(trifluoromethoxy)toluene
Formula:
C₈H₆BrF₃O Boiling Pt: 178…180 °C (1013 hPa) Density: 1.572 g/cm³ (20 °C) |
MDL Number:
MFCD00061271 CAS Number: 159689-88-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...