α-Bromo-2-(trifluoromethoxy)toluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

007978-1G 007978-5G FL7978.10 007978-25G
FLUO007978-1GEA 0 GBP
FLUO007978-1G FLUO007978-5G CHMPFL7978.10 FLUO007978-25G
α-Bromo-2-(trifluoromethoxy)toluene
α-Bromo-2-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 32 °C
Density: 1.583 g/cm³ (20 °C)
MDL Number: MFCD00236321
CAS Number: 198649-68-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR