α-Bromo-2-(difluoromethoxy)toluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

007861-5G 007861.25G 007861-1G
FLUO007861-5GEA 0 GBP
FLUO007861-5G FLUO007861.25G FLUO007861-1G
α-Bromo-2-(difluoromethoxy)toluene
α-Bromo-2-(difluoromethoxy)toluene
Formula: C₈H₇BrF₂O
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 13 °C
Density: 1.555 g/cm³ (20 °C)
MDL Number: MFCD00190103
CAS Number: 85684-64-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR