5-Bromo-α,α,α-trifluoro-2-nitrotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002247.10G 002247.50G 002247-25G 002247-100G
FLUO002247.10GEA 20.6 GBP
FLUO002247.10G FLUO002247.50G FLUO002247-25G FLUO002247-100G
5-Bromo-α,α,α-trifluoro-2-nitrotoluene
5-Bromo-α,α,α-trifluoro-2-nitrotoluene
Formula: C₇H₃BrF₃NO₂
Boiling Pt: 95…100 °C (5 mmHg)
Melting Pt: 40…45 °C
Density: 1.799 g/cm³ (25 °C)
MDL Number: MFCD00014706
CAS Number: 344-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR