2-Bromo-5-(trifluoromethyl)aniline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3-Amino-4-bromobenzotrifluoride, 6-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-4-bromo-α,α,α-trifluorotoluene, 3-Amino-4-brom-α,α,α-trifluortoluol
M-Clarity™ quality level = MQ100
217867-1G
217867-5G
SIAL217867-1GEA
45.6
GBP
SIAL217867-1G
SIAL217867-5G
2-Bromo-5-(trifluoromethyl)aniline, Sigma-Aldrich®
2-Bromo-5-(trifluoromethyl)aniline
Formula:
C₇H₅BrF₃N MW: 240.02 g/mol Boiling Pt: 94…96 °C (12 mmHg) Melting Pt: 47…49 °C Density: 1.694 g/cm³ (25 °C) |
MDL Number:
MFCD00007637 CAS Number: 454-79-5 EINECS: 207-227-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...