5-Bromo-α,α,α,2-tetrafluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-fluorobenzotrifluoride, 1-Bromo-4-fluoro-3-(trifluoromethyl)benzene

CFF578.5 004218-25G 004218-100G
CHMPCFF578.5EA 0 GBP
CHMPCFF578.5 FLUO004218-25G FLUO004218-100G
5-Bromo-α,α,α,2-tetrafluorotoluene
5-Bromo-α,α,α,2-tetrafluorotoluene
Formula: C₇H₃BrF₄
MDL Number: MFCD00042493
CAS Number: 393-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR