2-Bromo-α,α,α,4-tetrafluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-fluorobenzotrifluoride, 1-Bromo-3-fluoro-6-(trifluoromethyl)benzene

021893-1G 021893-5G
FLUO021893-1GEA 0 GBP
FLUO021893-1G FLUO021893-5G
2-Bromo-α,α,α,4-tetrafluorotoluene
2-Bromo-α,α,α,4-tetrafluorotoluene
Formula: C₇H₃BrF₄
MDL Number: MFCD03094041
CAS Number: 351003-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR