3-Bromo-5-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021361-5G 021361-25G
FLUO021361-5GEA 0 GBP
FLUO021361-5G FLUO021361-25G
3-Bromo-5-fluorotoluene
3-Bromo-5-fluorotoluene
Formula: C₇H₆BrF
MDL Number: MFCD01861195
CAS Number: 202865-83-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR