4-Bromo-3-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

002344.5G 002344-25G 002344-100G
FLUO002344.5GEA 20.6 GBP
FLUO002344.5G FLUO002344-25G FLUO002344-100G
4-Bromo-3-fluorotoluene
4-Bromo-3-fluorotoluene
Formula: C₇H₆BrF
Boiling Pt: 186 °C (1013 hPa)
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00040940
CAS Number: 452-74-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR