4-Bromo-α,α,α,3-tetrafluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

004217-5G 004217-25G
FLUO004217-5GEA 0 GBP
FLUO004217-5G FLUO004217-25G
4-Bromo-α,α,α,3-tetrafluorotoluene
4-Bromo-α,α,α,3-tetrafluorotoluene
Formula: C₇H₃BrF₄
MDL Number: MFCD00042497
CAS Number: 40161-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR