3,5-Difluorobenzyl bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

004798-10G 004798-100G 004798-25G
FLUO004798-10GEA 0 GBP
FLUO004798-10G FLUO004798-100G FLUO004798-25G
3,5-Difluorobenzyl bromide
3,5-Difluorobenzyl bromide
Formula: C₇H₅BrF₂
Boiling Pt: 65 °C (4.5 mmHg)
Density: 1.6 g/cm³ (20 °C)
MDL Number: MFCD00010304
CAS Number: 141776-91-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR