2,3-Difluorobenzyl bromide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
004796-5G
004796-25G
FLUO004796-5GEA
0
GBP
FLUO004796-5G
FLUO004796-25G
2,3-Difluorobenzyl bromide
2,3-Difluorobenzyl bromide
Formula:
C₇H₅BrF₂ Boiling Pt: 186 °C (1013 hPa) Melting Pt: 8 °C Density: 1.62 g/cm³ (20 °C) |
MDL Number:
MFCD00042488 CAS Number: 113211-94-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...