2,3,5-Trifluorobenzyl bromide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
α-Bromo-2,3,5-trifluorotoluene
007968-1G
007968-5G
007968-25G
FLUO007968-1GEA
0
GBP
FLUO007968-1G
FLUO007968-5G
FLUO007968-25G
2,3,5-Trifluorobenzyl bromide
2,3,5-Trifluorobenzyl bromide
Formula:
C₇H₄BrF₃ MW: 225.01 g/mol Boiling Pt: 182 °C (1013 hPa) Melting Pt: 21 °C |
MDL Number:
MFCD00083543 CAS Number: 226717-83-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...