2,3,4-Trifluorobenzyl bromide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1-(Bromomethyl)-2,3,4-trifluorobenzene, α-Bromo-2,3,4-trifluorotoluene
007503-1G
007503-5G
007503-10G
FLUO007503-1GEA
0
GBP
FLUO007503-1G
FLUO007503-5G
FLUO007503-10G
2,3,4-Trifluorobenzyl bromide
2,3,4-Trifluorobenzyl bromide
Formula:
C₇H₄BrF₃ MW: 225.01 g/mol Boiling Pt: 182 Melting Pt: 21 Density: 1.71 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00061233 CAS Number: 157911-55-2 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...