3-Bromo-o-toluic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018295-5G
018295-25G
FLUO018295-5GEA
0
GBP
FLUO018295-5G
FLUO018295-25G
3-Bromo-o-toluic acid
3-Bromo-o-toluic acid
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol |
MDL Number:
MFCD00270097 CAS Number: 76006-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...