ω-Bromo-2-methoxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methoxyphenacyl bromide, α-Bromo-2-methoxyacetophenone, 2-Bromo-2'-methoxyacetophenone, 2-Bromo-1-(2-methoxyphenyl)ethanone, 2-Bromo-1-(2-methoxyphenyl)ethan-1-one, Bromomethyl o-methoxyphenyl ketone
M-Clarity™ quality level = MQ100
100854-10G
100854-1G
100854-5G
SIAL100854-10GEA
219
GBP
SIAL100854-10G
SIAL100854-1G
SIAL100854-5G
ω-Bromo-2-methoxyacetophenone, Sigma-Aldrich®
ω-Bromo-2-methoxyacetophenone
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 324 °C (1013 hPa) Melting Pt: 40…44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000196 CAS Number: 31949-21-0 EINECS: 250-870-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...