3'-Bromoacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: m-Bromoacetophenone

008816-100G 008816-25G
FLUO008816-100GEA 0 GBP
FLUO008816-100G FLUO008816-25G
3'-Bromoacetophenone
3'-Bromoacetophenone
Formula: C₈H₇BrO
Boiling Pt: 255 °C (1013 hPa)
Density: 1,49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000083
CAS Number: 2142-63-4
EINECS: 218-396-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR