ω,3-Dibromoacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011106-10G 011106-100G
FLUO011106-10GEA 0 GBP
FLUO011106-10G FLUO011106-100G
ω,3-Dibromoacetophenone
ω,3-Dibromoacetophenone
Formula: C₈H₆Br₂O
Melting Pt: 51 °C
MDL Number: MFCD00082698
CAS Number: 18523-22-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR