α-Bromo-3-chloro-2-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-3-chloro-2-fluorobenzene

004445-1G 004445-5G 004445-25G 04445.25G
FLUO004445-1GEA 0 GBP
FLUO004445-1G FLUO004445-5G FLUO004445-25G FLUO04445.25G
α-Bromo-3-chloro-2-fluorotoluene
α-Bromo-3-chloro-2-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
Boiling Pt: 125…126 °C (15 mmHg)
MDL Number: MFCD00042503
CAS Number: 85070-47-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR