α-Bromo-2-chloro-5-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005631-1G 005631-5G
FLUO005631-1GEA 0 GBP
FLUO005631-1G FLUO005631-5G
α-Bromo-2-chloro-5-fluorotoluene
α-Bromo-2-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
MDL Number: MFCD01631419
CAS Number: 81778-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR