ω-Bromo-3-chloroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-3'-chloroacetophenone, Bromomethyl m-chlorophenyl ketone, 2-Bromo-1-(3-chlorophenyl)ethanone, 3-Chlorophenacyl bromide, α-Bromo-3-chloroacetophenone

CFF145.2.5 CFF145.10 017212-1G 017212-5G
CHMPCFF145.2.5EA 0 GBP
CHMPCFF145.2.5 CHMPCFF145.10 FLUO017212-1G FLUO017212-5G
ω-Bromo-3-chloroacetophenone
ω-Bromo-3-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00792669
CAS Number: 41011-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR